Learn More
Medchemexpress LLC D-Glucuronic acid lactone | 32449-92-6 | 98.0% | 176.12 | 1 ML
SDP
Catalog No. 5000405210
Encompass_Preferred
D-Glucuronic acid lactone, also known by various synonyms, is an endogenous metabolite and a derivative of glucuronic acid. This compound acts as a foundational reagent for various syntheses, making it suitable for a range of research applications.
  • Preparation of 1-O-acyl glucuronic acids for the anti-inflammatory agent mL-3000.
  • Synthesis of optically active glucuronic acids.
  • Production of long-chain alkyl glucuronides.
  • Potential utility in research pertaining to reversible cerebral vasoconstriction syndrome (RCVS).
Catalog No. 50-004-05210 Supplier Medchemexpress LLC Supplier No. HY4198210MM/1ML
D-Glucuronic acid lactone, also known by various synonyms, is an endogenous metabolite and a derivative of glucuronic acid. This compound acts as a foundational reagent for various syntheses, making it suitable for a range of research applications.
  • Preparation of 1-O-acyl glucuronic acids for the anti-inflammatory agent mL-3000.
  • Synthesis of optically active glucuronic acids.
  • Production of long-chain alkyl glucuronides.
  • Potential utility in research pertaining to reversible cerebral vasoconstriction syndrome (RCVS).