Learn More
Medchemexpress LLC D-Glucuronic acid lactone | 32449-92-6 | MFCD00135622 | 98.0% | 176.12 | 5 G
SDP
Catalog No. 5000404111
Encompass_Preferred
D-Glucuronic acid lactone is an endogenous metabolite and a glucuronic acid derivative. It serves as a starting reagent for the synthesis of 2,3,4-tris (tert-butyldimethylsilyl) glucuronic acid trichloroethyl ester, which is used to prepare 1-O-acyl glucuronic acids, synthesize optically active glucuronic acids, and produce long-chain alkyl glucuronides. It also shows potential for use in studies of reversible cerebral vasoconstriction syndrome (RCVS).
  • Endogenous metabolite and glucuronic acid derivative
  • Starting reagent for the synthesis of specific glucuronic acid esters
  • Prepares 1-O-acyl glucuronic acids
  • Synthesizes optically active glucuronic acids
  • Produces long-chain alkyl glucuronides
  • Potential use in studies of reversible cerebral vasoconstriction syndrome (RCVS)
Catalog No. 50-004-04111 Supplier Medchemexpress LLC Supplier No. HY419825G
D-Glucuronic acid lactone is an endogenous metabolite and a glucuronic acid derivative. It serves as a starting reagent for the synthesis of 2,3,4-tris (tert-butyldimethylsilyl) glucuronic acid trichloroethyl ester, which is used to prepare 1-O-acyl glucuronic acids, synthesize optically active glucuronic acids, and produce long-chain alkyl glucuronides. It also shows potential for use in studies of reversible cerebral vasoconstriction syndrome (RCVS).
  • Endogenous metabolite and glucuronic acid derivative
  • Starting reagent for the synthesis of specific glucuronic acid esters
  • Prepares 1-O-acyl glucuronic acids
  • Synthesizes optically active glucuronic acids
  • Produces long-chain alkyl glucuronides
  • Potential use in studies of reversible cerebral vasoconstriction syndrome (RCVS)