Learn More
Medchemexpress LLC D-Glucuronic acid (sodium) (monohydrate)-13C6 | 207300-70-7 | 25 MG
SDP
Catalog No. 5000421859
Encompass_Preferred
D-Glucuronic acid sodium monohydrate-13C6 is the 13C-labeled version of D-Glucuronic acid sodium salt monohydrate, an endogenous metabolite. This compound is suitable for various quantitative analysis applications.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • High purity of 99.9%
  • Appearance as a white to off-white solid
Catalog No. 50-004-21859 Supplier Medchemexpress LLC Supplier No. 25MGHYN6612S
D-Glucuronic acid sodium monohydrate-13C6 is the 13C-labeled version of D-Glucuronic acid sodium salt monohydrate, an endogenous metabolite. This compound is suitable for various quantitative analysis applications.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • High purity of 99.9%
  • Appearance as a white to off-white solid