Learn More
Medchemexpress LLC D-Glucuronic acid sodium salt monohydrate | 207300-70-7 | 1 G
SDP
Catalog No. 5000420140
Encompass_Preferred
D-Glucuronic acid sodium salt monohydrate is a primary constituent of numerous anti-inflammatory proteoglycans, facilitating embryonic development and inhibiting cell aggregation. When metabolized into ethyl glucuronide, it activates Toll-like receptor 4 (TLR4), leading to pain. Both D-Glucuronic acid sodium salt monohydrate and its derivative, glucurono-lactone, can function as liver detoxifiers for human health prevention, and its derivatives also exhibit anti-tumor activity. It is intended for research use only.
  • Major component of many anti-inflammatory proteoglycans.
  • Can promote embryonic development.
  • Inhibits cell aggregation.
  • Activates Toll-like receptor 4 (TLR4) after metabolism into ethyl glucuronide, causing pain.
  • Serves as a liver detoxifier for human health prevention.
  • Derivatives possess anti-tumor activity.
Catalog No. 50-004-20140 Supplier Medchemexpress LLC Supplier No. HYN6612A1G
D-Glucuronic acid sodium salt monohydrate is a primary constituent of numerous anti-inflammatory proteoglycans, facilitating embryonic development and inhibiting cell aggregation. When metabolized into ethyl glucuronide, it activates Toll-like receptor 4 (TLR4), leading to pain. Both D-Glucuronic acid sodium salt monohydrate and its derivative, glucurono-lactone, can function as liver detoxifiers for human health prevention, and its derivatives also exhibit anti-tumor activity. It is intended for research use only.
  • Major component of many anti-inflammatory proteoglycans.
  • Can promote embryonic development.
  • Inhibits cell aggregation.
  • Activates Toll-like receptor 4 (TLR4) after metabolism into ethyl glucuronide, causing pain.
  • Serves as a liver detoxifier for human health prevention.
  • Derivatives possess anti-tumor activity.