Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-Glucuronic acid sodium salt monohydrate | 207300-70-7 | 10 G
SDP

Catalog No. 5000417992
Encompass_Preferred

D-Glucuronic acid sodium salt monohydrate is a primary component of several anti-inflammatory proteoglycans, which can foster embryonic development and hinder cell aggregation. When metabolized into ethyl glucuronide, it activates Toll-like receptor 4 (TLR4), leading to pain. This compound and its derivative, glucurono-lactone, can act as liver detoxifiers for human health prevention, with its derivatives also exhibiting anti-tumor activity.

  • Purity: 98.0%
  • Molecular weight: 234.14
  • Formula: C6H11NaO8
  • Appearance: Solid, white to off-white
  • Acts as a component of anti-inflammatory proteoglycans
  • Promotes early embryonic development
  • Inhibits cell aggregation
  • Functions as a liver detoxifier
  • Derivatives show anti-tumor activity
  • Activates Toll-like receptor 4 (TLR4) when metabolized
  • Categorized as a ketone, aldehyde, and acid

Catalog No. 50-004-17992 Supplier Medchemexpress LLC Supplier No. HYN6612A10G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Glucuronic acid sodium salt monohydrate is a primary component of several anti-inflammatory proteoglycans, which can foster embryonic development and hinder cell aggregation. When metabolized into ethyl glucuronide, it activates Toll-like receptor 4 (TLR4), leading to pain. This compound and its derivative, glucurono-lactone, can act as liver detoxifiers for human health prevention, with its derivatives also exhibiting anti-tumor activity.

  • Purity: 98.0%
  • Molecular weight: 234.14
  • Formula: C6H11NaO8
  • Appearance: Solid, white to off-white
  • Acts as a component of anti-inflammatory proteoglycans
  • Promotes early embryonic development
  • Inhibits cell aggregation
  • Functions as a liver detoxifier
  • Derivatives show anti-tumor activity
  • Activates Toll-like receptor 4 (TLR4) when metabolized
  • Categorized as a ketone, aldehyde, and acid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.