Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Glucuronic Acid, Spectrum™ Chemical
SDP

Synonym: D-Glucopyranuronate, D-Glucopyranuronic Acid, D-Glucuronate

Supplier:  Spectrum Chemical Manufacturing Corporation G2028100GM

Quantity 100 g

 View more versions of this product

Catalog No. 18-620-777


Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart

Description

Description

D-Glucuronic Acid, also known as β-D-glucopyranuronic acid, is a carboxylic acid with a structure similar to glucose and used in the determination of urinary steroids and of steroid conjugates in blood. Ungraded products supplied by Spectrum™ are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Specifications

Chemical Identifiers

6556-12-3
194.14 g/mol

Specifications

6556-12-3
Amber Glass Bottle
D-Glucuronic Acid
194.14 g/mol
100 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Promo
Expires: 06-30-2027
Get Up to $500 in Chemicals with Purchase
Spend $250 or more on Spectrum Chemical chemicals and get up to $500 to spend on additional chemicals.

Adding...

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.