Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Glucuronic Acid, Spectrum™ Chemical
SDP

Synonym: D-Glucopyranuronate, D-Glucopyranuronic Acid, D-Glucuronate

$127.00 - $9944.00

Chemical Identifiers

CAS 6556-12-3
Molecular Weight (g/mol) 194.14 g/mol
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
18-620-775
SDP
View Documents
Spectrum Chemical Manufacturing Corporation
G20285GM
5 g
Each for $127.00
Add to Cart
 
18-620-776
SDP
View Documents
Spectrum Chemical Manufacturing Corporation
G202825GM
25 g
Each for $288.00
Add to Cart
 
18-620-777
SDP
View Documents
Spectrum Chemical Manufacturing Corporation
G2028100GM
100 g
Each for $923.00
Add to Cart
 
18-620-778
SDP
View Documents
Spectrum Chemical Manufacturing Corporation
G20282KGBL
2 kg
Each for $9,944.00
Add to Cart
 
Description

Description

D-Glucuronic Acid, also known as β-D-glucopyranuronic acid, is a carboxylic acid with a structure similar to glucose and used in the determination of urinary steroids and of steroid conjugates in blood. Ungraded products supplied by Spectrum™ are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Specifications

Chemical Identifiers

6556-12-3
194.14 g/mol

Specifications

6556-12-3
194.14 g/mol
Amber Glass Bottle
D-Glucuronic Acid
Videos
SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.