Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Glucuronic Acid, Spectrum™ Chemical
SDP

Catalog No. 18620778 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
2 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-778 2 kg
18-620-775 5 g
18-620-776 25 g
18-620-777 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-620-778 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. G20282KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Synonym: D-Glucopyranuronate, D-Glucopyranuronic Acid, D-Glucuronate

D-Glucuronic Acid, also known as β-D-glucopyranuronic acid, is a carboxylic acid with a structure similar to glucose and used in the determination of urinary steroids and of steroid conjugates in blood. Ungraded products supplied by Spectrum™ are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Chemical Identifiers

CAS 6556-12-3
Molecular Weight (g/mol) 194.14 g/mol

Specifications

Packaging Amber Glass Bottle
Quantity 2 kg
Chemical Name or Material D-Glucuronic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.