Learn More
Medchemexpress LLC D-Glucuronic acid (Standard) | 6556-12-3 | 100 MG
SDP
Catalog No. 5000417873
Encompass_Preferred
D-Glucuronic acid (Standard) is an analytical standard intended for research and analytical applications. It is a significant component of numerous anti-inflammatory proteoglycans, contributing to embryonic development and inhibiting cell aggregation. When metabolized into ethyl glucuronide, it activates Toll-like receptor 4 (TLR4), which is associated with pain. Furthermore, D-Glucuronic acid and its derivative, glucurono-lactone, are known as liver detoxifiers and its derivatives also exhibit anti-tumor activity.
  • Analytical standard for assays
  • Intended for research and analytical use
  • Utilized in HPLC, GC, and MS research experiments
  • Major component of anti-inflammatory proteoglycans
  • Promotes embryonic development
  • Inhibits cell aggregation
  • Acts as a liver detoxifier
  • Derivatives possess anti-tumor activity
  • Solid, white to off-white appearance
  • Purity of 99.93%
Catalog No. 50-004-17873 Supplier Medchemexpress LLC Supplier No. HYN6612R100MG
D-Glucuronic acid (Standard) is an analytical standard intended for research and analytical applications. It is a significant component of numerous anti-inflammatory proteoglycans, contributing to embryonic development and inhibiting cell aggregation. When metabolized into ethyl glucuronide, it activates Toll-like receptor 4 (TLR4), which is associated with pain. Furthermore, D-Glucuronic acid and its derivative, glucurono-lactone, are known as liver detoxifiers and its derivatives also exhibit anti-tumor activity.
  • Analytical standard for assays
  • Intended for research and analytical use
  • Utilized in HPLC, GC, and MS research experiments
  • Major component of anti-inflammatory proteoglycans
  • Promotes embryonic development
  • Inhibits cell aggregation
  • Acts as a liver detoxifier
  • Derivatives possess anti-tumor activity
  • Solid, white to off-white appearance
  • Purity of 99.93%