Learn More
Medchemexpress LLC D-Glucuronic acid (Standard) | 6556-12-3 | 25 G
SDP
Catalog No. 5000421975
Encompass_Preferred
D-Glucuronic acid (Standard) is the analytical standard of D-Glucuronic acid, intended for research and analytical applications. It is a major component of many anti-inflammatory proteoglycans, which can promote embryonic development and inhibit cell aggregation. After being metabolized into ethyl glucuronide, D-Glucuronic acid activates Toll-like receptor 4 (TLR4), causing pain. D-Glucuronic acid and its derivative glucurono-lactone can serve as liver detoxifiers for human health prevention, and its derivatives also possess anti-tumor activity.
  • Analytical standard for D-Glucuronic acid
  • Intended for research and analytical applications
  • Promotes embryonic development and inhibits cell aggregation
  • Activates Toll-like receptor 4 (TLR4) when metabolized
  • Serves as a liver detoxifier; derivatives possess anti-tumor activity
  • Commonly used in HPLC, GC, and MS for research experiments
Catalog No. 50-004-21975 Supplier Medchemexpress LLC Supplier No. 25GHYN6612A
D-Glucuronic acid (Standard) is the analytical standard of D-Glucuronic acid, intended for research and analytical applications. It is a major component of many anti-inflammatory proteoglycans, which can promote embryonic development and inhibit cell aggregation. After being metabolized into ethyl glucuronide, D-Glucuronic acid activates Toll-like receptor 4 (TLR4), causing pain. D-Glucuronic acid and its derivative glucurono-lactone can serve as liver detoxifiers for human health prevention, and its derivatives also possess anti-tumor activity.
  • Analytical standard for D-Glucuronic acid
  • Intended for research and analytical applications
  • Promotes embryonic development and inhibits cell aggregation
  • Activates Toll-like receptor 4 (TLR4) when metabolized
  • Serves as a liver detoxifier; derivatives possess anti-tumor activity
  • Commonly used in HPLC, GC, and MS for research experiments