Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-Glucuronic acid (standard) | 6556-12-3 | 250 MG
SDP

Catalog No. 5000420695
Encompass_Preferred

D-Glucuronic acid (Standard) is an analytical standard for research and analytical applications. It is a key component of anti-inflammatory proteoglycans, promoting embryonic development and inhibiting cell aggregation. Metabolized into ethyl glucuronide, it activates Toll-like receptor 4 (TLR4), potentially causing pain. This compound and its derivative glucurono-lactone act as liver detoxifiers, with derivatives also showing anti-tumor activity. It is utilized in qualitative, quantitative, and methodological research, particularly with techniques like HPLC, GC, and MS.

  • Purity: 99.93%
  • Molecular weight: 194.14
  • Chemical formula: C6H10O7
  • Appearance: Solid, white to off-white
  • Promotes embryonic development
  • Liver detoxifier

Catalog No. 50-004-20695 Supplier Medchemexpress LLC Supplier No. 250MGHYN6612R
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Glucuronic acid (Standard) is an analytical standard for research and analytical applications. It is a key component of anti-inflammatory proteoglycans, promoting embryonic development and inhibiting cell aggregation. Metabolized into ethyl glucuronide, it activates Toll-like receptor 4 (TLR4), potentially causing pain. This compound and its derivative glucurono-lactone act as liver detoxifiers, with derivatives also showing anti-tumor activity. It is utilized in qualitative, quantitative, and methodological research, particularly with techniques like HPLC, GC, and MS.

  • Purity: 99.93%
  • Molecular weight: 194.14
  • Chemical formula: C6H10O7
  • Appearance: Solid, white to off-white
  • Promotes embryonic development
  • Liver detoxifier

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.