Learn More
Medchemexpress LLC D-Glucuronic acid (Standard) | 6556-12-3 | 50 MG
SDP
Catalog No. 5000420139
Encompass_Preferred
D-Glucuronic acid (Standard) is the analytical standard of D-Glucuronic acid, intended for research and analytical applications. It is a major component of many anti-inflammatory proteoglycans, promoting embryonic development and inhibiting cell aggregation. This compound and its derivative glucurono-lactone can also serve as liver detoxifiers for human health prevention, and its derivatives possess anti-tumor activity.
  • Analytical standard for research and analytical applications
  • Major component of many anti-inflammatory proteoglycans
  • Promotes embryonic development and inhibits cell aggregation
  • Can serve as a liver detoxifier
  • Derivatives possess anti-tumor activity
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
Catalog No. 50-004-20139 Supplier Medchemexpress LLC Supplier No. HYN6612R50MG
D-Glucuronic acid (Standard) is the analytical standard of D-Glucuronic acid, intended for research and analytical applications. It is a major component of many anti-inflammatory proteoglycans, promoting embryonic development and inhibiting cell aggregation. This compound and its derivative glucurono-lactone can also serve as liver detoxifiers for human health prevention, and its derivatives possess anti-tumor activity.
  • Analytical standard for research and analytical applications
  • Major component of many anti-inflammatory proteoglycans
  • Promotes embryonic development and inhibits cell aggregation
  • Can serve as a liver detoxifier
  • Derivatives possess anti-tumor activity
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS