Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC β-D-glucuronide-pNP-carbonate | 894095-98-8 | MFCD31715405 | 99.6% | 913.83 | C45H43N3O18 | 250 MG
SDP

Catalog No. 5000203950
Encompass_Preferred

β-D-glucuronide-pNP-carbonate is a cleavable glucuronide-based carbonate linker used as a precursor in antibody-drug conjugate (ADC) synthesis. It features a p-nitrophenyl (pNP) activated carbonate that facilitates conjugation and enzymatic payload release upon glucuronidase cleavage. Supplied as a white to off-white solid with high purity and defined molecular properties, it is intended for research applications.

  • Cleavable glucuronide-based ADC linker for payload release
  • Activated p-nitrophenyl carbonate handle for efficient conjugation
  • High purity (99.6%) suitable for research applications
  • Molecular weight 913.83 and formula C45H43N3O18
  • Store at -20°C, sealed and protected from moisture

Catalog No. 50-002-03950 Supplier Medchemexpress LLC Supplier No. HY136329250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

β-D-glucuronide-pNP-carbonate is a cleavable glucuronide-based carbonate linker used as a precursor in antibody-drug conjugate (ADC) synthesis. It features a p-nitrophenyl (pNP) activated carbonate that facilitates conjugation and enzymatic payload release upon glucuronidase cleavage. Supplied as a white to off-white solid with high purity and defined molecular properties, it is intended for research applications.

  • Cleavable glucuronide-based ADC linker for payload release
  • Activated p-nitrophenyl carbonate handle for efficient conjugation
  • High purity (99.6%) suitable for research applications
  • Molecular weight 913.83 and formula C45H43N3O18
  • Store at -20°C, sealed and protected from moisture

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.