Learn More
Medchemexpress LLC D-Glucurono-6,3-lactone acetonide | 20513-98-8 | >98% | 216.19 | 50 G
SDP
Catalog No. 5000403471
Encompass_Preferred
D-Glucurono-6,3-lactone acetonide can be converted to optically active and partially protected inositols. This product is for research use only and not intended for patient use.
  • For research use only
  • Can be converted to optically active and partially protected inositols
Catalog No. 50-004-03471 Supplier Medchemexpress LLC Supplier No. HY4184050G
D-Glucurono-6,3-lactone acetonide can be converted to optically active and partially protected inositols. This product is for research use only and not intended for patient use.
  • For research use only
  • Can be converted to optically active and partially protected inositols