Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Glucurono-6,3-lactone acetonide, 98+%

Catalog No. AAL1776509
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1776509 10 g
AAL1776518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1776509 Supplier Thermo Scientific Chemicals Supplier No. L1776509
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 20513-98-8 | C9H12O6 | 216.189 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20513-98-8
Molecular Formula C9H12O6
Molecular Weight (g/mol) 216.189
MDL Number MFCD00061641
InChI Key BDBGJSXZKMTMGP-BOFVIFSBSA-N
Synonym d-glucurono-6,3-lactone acetonide
PubChem CID 134129430
IUPAC Name (3aS,3bS,6S,6aS,7aR)-6-hydroxy-2,2-dimethyl-3b,6,6a,7a-tetrahydro-3aH-furo[2,3]furo[2,4-b][1,3]dioxol-5-one
SMILES CC1(OC2C3C(C(C(=O)O3)O)OC2O1)C

Specifications

Melting Point 119°C to 121°C
Quantity 10 g
Optical Rotation +52° (c=1 in Chloroform)
Formula Weight 216.19
Percent Purity ≥98%
Chemical Name or Material D-Glucurono-6,3-lactone acetonide
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.