Learn More
Cayman Chemical D-Glutamic Acid, Purity: ≥95%, CAS: 6893-26-1, 100 g
SDP
Catalog No. 502991854
Encompass_Preferred
An amino acid and a component of bacterial peptidoglycan; is also a component of poly-γ-glutamic acid
Catalog No. 50-299-1854 Supplier Cayman Chemical Supplier No. 31503100 G
An amino acid and a component of bacterial peptidoglycan; is also a component of poly-γ-glutamic acid