Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-glutamic acid | 6893-26-1 | MFCD00063112 | ≥99.0% | 147.13 | C5H9NO4 | 50 G
SDP

Catalog No. 5000211962
Encompass_Preferred

D-glutamic acid is the D-enantiomer of glutamic acid supplied as a research-grade analytical standard. It is used in pharmaceutical, food, and biochemical research applications where stereospecific amino acid reference material is required. The material is provided as a dry powder and intended for research use only.

  • High purity ≥99.0%.
  • Molecular formula C5H9NO4.
  • Molecular weight 147.13 g/mol.
  • Available in laboratory pack sizes, including 50 G.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (2 years) or -20°C (1 year).
  • Intended for research use only.

Catalog No. 50-002-11962 Supplier Medchemexpress LLC Supplier No. HY10080550G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-glutamic acid is the D-enantiomer of glutamic acid supplied as a research-grade analytical standard. It is used in pharmaceutical, food, and biochemical research applications where stereospecific amino acid reference material is required. The material is provided as a dry powder and intended for research use only.

  • High purity ≥99.0%.
  • Molecular formula C5H9NO4.
  • Molecular weight 147.13 g/mol.
  • Available in laboratory pack sizes, including 50 G.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (2 years) or -20°C (1 year).
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.