Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-glutamic acid | 6893-26-1 | MFCD00063112 | >99.0% | 147.13 g/mol | C5H9NO4 | 5 G
SDP

Catalog No. 5000212965
Encompass_Preferred

D-glutamic acid is the D-enantiomer of L-glutamic acid and is used as an analytical standard and research reagent in pharmaceutical, biochemical, and food-related applications. It is supplied in small pack sizes suitable for laboratory use and analytical testing.

  • High chemical purity suitable for analytical use.
  • Applicable as an analytical standard and research reagent.
  • Used in pharmaceutical and food-related research.
  • Available in small pack sizes for laboratory experiments.
  • Molecular weight 147.13 g/mol; water-soluble amino acid.
  • CAS number 6893-26-1 for unambiguous identification.

Catalog No. 50-002-12965 Supplier Medchemexpress LLC Supplier No. HY1008055G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-glutamic acid is the D-enantiomer of L-glutamic acid and is used as an analytical standard and research reagent in pharmaceutical, biochemical, and food-related applications. It is supplied in small pack sizes suitable for laboratory use and analytical testing.

  • High chemical purity suitable for analytical use.
  • Applicable as an analytical standard and research reagent.
  • Used in pharmaceutical and food-related research.
  • Available in small pack sizes for laboratory experiments.
  • Molecular weight 147.13 g/mol; water-soluble amino acid.
  • CAS number 6893-26-1 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.