Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D(-)-Glutamic acid, 99+%

Catalog No. p-138096
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC227890050 5 g Glass bottle
AC227890250 25 g Glass bottle
AC227891000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC227890050 Supplier Thermo Scientific Chemicals Supplier No. 227890050
Only null left
Add to Cart

CAS: 6893-26-1 | C5H9NO4 | 147.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6893-26-1
Molecular Weight (g/mol) 147.13
MDL Number MFCD00063112
InChI Key WHUUTDBJXJRKMK-GSVOUGTGSA-N
Synonym d-glutamic acid, h-d-glu-oh, d-glu, r-2-aminopentanedioic acid, 2r-2-aminopentanedioic acid, d-glutamate, d-2-aminopentanedioic acid, glutamic acid d-form, d--glutamic acid, d-glutaminic acid
PubChem CID 23327
ChEBI CHEBI:15966
IUPAC Name (2R)-2-aminopentanedioic acid
SMILES C(CC(=O)O)C(C(=O)O)N

Specifications

Melting Point 200.0°C to 202.0°C
Color White
Loss on Drying 0.2% max. (105°C, 3 hrs)
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Molecular Formula C5H9NO4
Linear Formula HO2CCH2CH2CH(NH2)CO2H
Quantity 5 g
Merck Index 15, 4505
Specific Rotation Condition − 31.30 (20.00°C c=10,2 N HCl)
Specific Rotation − 31.30
Solubility Information Solubility in water: 7g/L (20°C). Other solubilities: practically insoluble in organic solvents
Formula Weight 147.13
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material D(-)-Glutamic acid, 99+%
Show More Show Less
EINECSNumber 230-000-8
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.