Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals D-Glutamic acid, 99+%

Catalog No. AAA1419114
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1419114 25 g
AAA1419106 5 g
AAA1419122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1419114 Supplier Thermo Scientific Chemicals Supplier No. A1419114
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 6893-26-1 | C5H9NO4 | 147.13 g/mol

D-Glutamic acid acts as a superior to alpha ketoglutarate. It helps in the growth of Lactobacillus arabinosus. It can inhibit the IgE binding to peanut allergens. It is widely used in pharmaceuticals and foods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Glutamic acid acts as a superior to alpha ketoglutarate. It helps in the growth of Lactobacillus arabinosus. It can inhibit the IgE binding to peanut allergens. It is widely used in pharmaceuticals and foods.

Solubility
Soluble in water (8.88 mg/mL) at 25°C.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6893-26-1
Molecular Formula C5H9NO4
Molecular Weight (g/mol) 147.13
MDL Number MFCD00063112
InChI Key WHUUTDBJXJRKMK-GSVOUGTGSA-N
Synonym d-glutamic acid, h-d-glu-oh, d-glu, r-2-aminopentanedioic acid, 2r-2-aminopentanedioic acid, d-glutamate, d-2-aminopentanedioic acid, glutamic acid d-form, d--glutamic acid, d-glutaminic acid
PubChem CID 23327
ChEBI CHEBI:15966
IUPAC Name (2R)-2-aminopentanedioic acid
SMILES C(CC(=O)O)C(C(=O)O)N

Specifications

Melting Point 199°C (decomposition)
Beilstein 1723800
Merck Index 14,4469
Solubility Information Soluble in water (8.88mg/mL) at 25°C.
Quantity 25 g
Formula Weight 147.13
Optical Rotation −30° (c=10 in 2N HCl)
Percent Purity ≥99%
Chemical Name or Material D-Glutamic acid
EINECSNumber 230-000-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.