Learn More
Cayman Chemical D-Glutamic Acid-d5, Purity: ≥99% deuterated forms (d1-d5), CAS: 14341-88-9, 10 mg
SDP
Catalog No. 502913111
Encompass_Preferred
An internal standard for the quantification of D-glutamic acid by GC- or LC-M
Catalog No. 50-291-3111 Supplier Cayman Chemical Supplier No. 3484010 MG
An internal standard for the quantification of D-glutamic acid by GC- or LC-M