Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-Glutamic acid, N-[(phenylmethoxy)carbonyl]-, 1-(phenylmethyl) ester | 65706-99-2 | MFCD00069647 | 99.5% | 371.38 | 10 G
SDP

Catalog No. 5000453867
Encompass_Preferred

Z-D-Glu-OBzl, also known by synonyms such as Cbz-D-Glu-Obzl and N-Cbz-D-glutamic acid alpha-benzyl ester, is an amino acid derivative with a molecular formula of C20H21NO6 and a molecular weight of 371.38. This compound is typically a white to off-white solid with a high purity of 99.54% (HPLC). It is intended for research use only.

  • Molecular weight: 371.38
  • Purity (HPLC): 99.54%
  • Appearance: White to off-white solid
  • Storage (Powder): -20°C for 3 years, 4°C for 2 years
  • Storage (In solvent): -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-53867 Supplier Medchemexpress LLC Supplier No. HYW01107510G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Z-D-Glu-OBzl, also known by synonyms such as Cbz-D-Glu-Obzl and N-Cbz-D-glutamic acid alpha-benzyl ester, is an amino acid derivative with a molecular formula of C20H21NO6 and a molecular weight of 371.38. This compound is typically a white to off-white solid with a high purity of 99.54% (HPLC). It is intended for research use only.

  • Molecular weight: 371.38
  • Purity (HPLC): 99.54%
  • Appearance: White to off-white solid
  • Storage (Powder): -20°C for 3 years, 4°C for 2 years
  • Storage (In solvent): -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.