Learn More
Selleck Chemical LLC D-glutamine 10mg 5959-95-5
SDP
Supplier:  Selleck Chemical LLC S189310MG
Encompass_Preferred
D-glutamine is a D type stereoisomer of glutamine which is one of the 20 amino acids encoded by the standard genetic code. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-756-2