Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-γ-glutamyl-D-glutamic acid | 4553-17-7 | 98.9% | 276.24 g/mol | C10H16N2O7 | 5 MG
SDP

Catalog No. 5000227787
Encompass_Preferred

D-γ-Glutamyl-D-glutamic acid is a γ-linked dipeptide of D-γ-glutamyl and D-glutamic acid residues supplied as a powdered research reagent (5 mg) with high purity for analytical and peptide studies.

  • High purity (~98.9%) suitable for analytical work.
  • Molecular formula C10H16N2O7; molecular weight 276.24 g/mol.
  • Provided as a lyophilized powder for stability and convenience.
  • Storage: sealed, away from moisture; powder -80°C (2 years) or -20°C (1 year).
  • Useful as an isomeric control and for structure-activity relationship studies.

Catalog No. 50-002-27787 Supplier Medchemexpress LLC Supplier No. HY118090A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-γ-Glutamyl-D-glutamic acid is a γ-linked dipeptide of D-γ-glutamyl and D-glutamic acid residues supplied as a powdered research reagent (5 mg) with high purity for analytical and peptide studies.

  • High purity (~98.9%) suitable for analytical work.
  • Molecular formula C10H16N2O7; molecular weight 276.24 g/mol.
  • Provided as a lyophilized powder for stability and convenience.
  • Storage: sealed, away from moisture; powder -80°C (2 years) or -20°C (1 year).
  • Useful as an isomeric control and for structure-activity relationship studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.