Learn More
Medchemexpress LLC γ-D-glutamylglycine | 6729-55-1 | MFCD00063109 | 99.2% | 204.18 g/mol | C7H12N2O5 | 1 ML
SDP
Catalog No. 5000213551
Encompass_Preferred
gamma-DGG (γ-D-glutamylglycine) is a competitive AMPA receptor blocker supplied as a 10 mM aqueous solution (1 mL). It reversibly reduces AMPA receptor-mediated synaptic currents and is commonly used in electrophysiology and neuropharmacology studies.
  • Competitive AMPA receptor blocker
  • Reversible inhibition of AMPA receptor-mediated currents
  • Provided as a 10 mM aqueous solution, 1 mL
  • High purity (99.16% by HPLC)
  • Molecular weight 204.18 g/mol
  • Suitable for electrophysiology and synaptic physiology studies
Catalog No. 50-002-13551 Supplier Medchemexpress LLC Supplier No. HY10078510MM 1ML
gamma-DGG (γ-D-glutamylglycine) is a competitive AMPA receptor blocker supplied as a 10 mM aqueous solution (1 mL). It reversibly reduces AMPA receptor-mediated synaptic currents and is commonly used in electrophysiology and neuropharmacology studies.
  • Competitive AMPA receptor blocker
  • Reversible inhibition of AMPA receptor-mediated currents
  • Provided as a 10 mM aqueous solution, 1 mL
  • High purity (99.16% by HPLC)
  • Molecular weight 204.18 g/mol
  • Suitable for electrophysiology and synaptic physiology studies