Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-glyceraldehyde 3-phosphate | 591-57-1 | MFCD00057581 | 99.8% | 170.06 g/mol | C3H7O6P | 10 MG
SDP

Catalog No. 5000287548
Encompass_Preferred

D-Glyceraldehyde 3-phosphate is an endogenous glycolytic intermediate commonly used in biochemical and metabolic research. The product is provided as a prepared aqueous solution with analytical documentation and storage guidance for laboratory use.

  • Endogenous metabolite relevant to glycolysis and gluconeogenesis.
  • Provided as an aqueous solution at 58.80 mM (1 mL) for convenient use.
  • High purity: 99.84%.
  • Molecular weight 170.06 g/mol and formula C3H7O6P.
  • Documentation available: SDS, COA, and RP-HPLC reports.
  • Store frozen (solution, -20°C) with a 2-year shelf life when stored as recommended.

Catalog No. 50-002-87548 Supplier Medchemexpress LLC Supplier No. HY15122310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Glyceraldehyde 3-phosphate is an endogenous glycolytic intermediate commonly used in biochemical and metabolic research. The product is provided as a prepared aqueous solution with analytical documentation and storage guidance for laboratory use.

  • Endogenous metabolite relevant to glycolysis and gluconeogenesis.
  • Provided as an aqueous solution at 58.80 mM (1 mL) for convenient use.
  • High purity: 99.84%.
  • Molecular weight 170.06 g/mol and formula C3H7O6P.
  • Documentation available: SDS, COA, and RP-HPLC reports.
  • Store frozen (solution, -20°C) with a 2-year shelf life when stored as recommended.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.