Learn More
Medchemexpress LLC D-glyceric acid sodium | 109175-64-6 | 99.0% | C3H5NaO4 | 10MG
SDP
Catalog No. 5000260245
Encompass_Preferred
D-glyceric acid sodium is the monosodium salt of D-glyceric acid, an endogenous metabolite supplied as a research-grade reagent for biochemical and metabolomics studies. It is commonly used in investigations of metabolic disorders and analytical method development where a characterized standard is required.
  • High purity (99.0%) suitable for analytical and research applications.
  • Monosodium salt form increases solubility in aqueous solutions.
  • Relevant for research into metabolic disorders such as primary hyperoxaluria type I and glutaric acidemia type II.
  • Solid, white to off-white powder that is stable under recommended storage conditions.
  • Available in small laboratory pack sizes for convenient use in assay development.
  • Documentation available including datasheet, certificate of analysis, and safety data sheet.
Catalog No. 50-002-60245 Supplier Medchemexpress LLC Supplier No. HY139070A10MG
D-glyceric acid sodium is the monosodium salt of D-glyceric acid, an endogenous metabolite supplied as a research-grade reagent for biochemical and metabolomics studies. It is commonly used in investigations of metabolic disorders and analytical method development where a characterized standard is required.
  • High purity (99.0%) suitable for analytical and research applications.
  • Monosodium salt form increases solubility in aqueous solutions.
  • Relevant for research into metabolic disorders such as primary hyperoxaluria type I and glutaric acidemia type II.
  • Solid, white to off-white powder that is stable under recommended storage conditions.
  • Available in small laboratory pack sizes for convenient use in assay development.
  • Documentation available including datasheet, certificate of analysis, and safety data sheet.