Learn More
Medchemexpress LLC D-GsMTx4 | 1209500-46-8 | 99.4% | 4095.84 | 1 MG
SDP
Catalog No. 5000423082
Encompass_Preferred
D-GsMTx4 is a spider peptide that serves as the D enantiomer of GsMTx4. This compound is a potent inhibitor of the mechanosensitive ion channel Piezo2, making it a valuable tool in neuroscience research. It has demonstrated inhibitory effects on intracellular calcium ([Ca2+]i) elevation, as well as the mTOR and PI3K-Akt signaling pathways. Research indicates its potential in mitigating mechanical allodynia and thermal hyperalgesia. D-GsMTx4 is utilized in studies exploring mechanical stress, chronic pain, and idiopathic pulmonary fibrosis.
  • Inhibits mechanosensitive ion channel Piezo2
  • Inhibits [Ca2+]i elevation
  • Inhibits mTOR and PI3K-Akt signaling pathways
  • Potentially mitigates mechanical allodynia and thermal hyperalgesia
  • Used in research of mechanical stress, chronic pain, and idiopathic pulmonary fibrosis
Catalog No. 50-004-23082 Supplier Medchemexpress LLC Supplier No. HYP1410B1MG
D-GsMTx4 is a spider peptide that serves as the D enantiomer of GsMTx4. This compound is a potent inhibitor of the mechanosensitive ion channel Piezo2, making it a valuable tool in neuroscience research. It has demonstrated inhibitory effects on intracellular calcium ([Ca2+]i) elevation, as well as the mTOR and PI3K-Akt signaling pathways. Research indicates its potential in mitigating mechanical allodynia and thermal hyperalgesia. D-GsMTx4 is utilized in studies exploring mechanical stress, chronic pain, and idiopathic pulmonary fibrosis.
  • Inhibits mechanosensitive ion channel Piezo2
  • Inhibits [Ca2+]i elevation
  • Inhibits mTOR and PI3K-Akt signaling pathways
  • Potentially mitigates mechanical allodynia and thermal hyperalgesia
  • Used in research of mechanical stress, chronic pain, and idiopathic pulmonary fibrosis