Learn More
Medchemexpress LLC D-GsMTx4 | 99.4% | 4095.84 | 100 UG
SDP
Catalog No. 5000424683
Encompass_Preferred
D-GsMTx4 is a spider peptide and the D enantiomer of GsMTx4. It acts as an inhibitor of the mechanosensitive ion channel Piezo2. It inhibits [Ca2+]i elevation and also impacts the mTOR and PI3K-Akt signaling pathways. D-GsMTx4 has been shown to inhibit mechanical allodynia and thermal hyperalgesia. It is suitable for research in areas such as mechanical stress, chronic pain, and idiopathic pulmonary fibrosis.
  • Inhibits the mechanosensitive ion channel Piezo2.
  • Inhibits [Ca2+]i elevation.
  • Inhibits mTOR and PI3K-Akt signaling pathways.
  • Inhibits mechanical allodynia and thermal hyperalgesia.
  • Suitable for research on mechanical stress, chronic pain, and idiopathic pulmonary fibrosis.
  • Decreases phosphorylation levels of proteins in the mTOR and PI3K-Akt signaling pathways in primary human lung fibroblasts.
Catalog No. 50-004-24683 Supplier Medchemexpress LLC Supplier No. HYP1410B100UG
D-GsMTx4 is a spider peptide and the D enantiomer of GsMTx4. It acts as an inhibitor of the mechanosensitive ion channel Piezo2. It inhibits [Ca2+]i elevation and also impacts the mTOR and PI3K-Akt signaling pathways. D-GsMTx4 has been shown to inhibit mechanical allodynia and thermal hyperalgesia. It is suitable for research in areas such as mechanical stress, chronic pain, and idiopathic pulmonary fibrosis.
  • Inhibits the mechanosensitive ion channel Piezo2.
  • Inhibits [Ca2+]i elevation.
  • Inhibits mTOR and PI3K-Akt signaling pathways.
  • Inhibits mechanical allodynia and thermal hyperalgesia.
  • Suitable for research on mechanical stress, chronic pain, and idiopathic pulmonary fibrosis.
  • Decreases phosphorylation levels of proteins in the mTOR and PI3K-Akt signaling pathways in primary human lung fibroblasts.