Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ D-GsMTx4
GREENER_CHOICE

Catalog No. 7170100U
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
7170100U 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 7170100U Supplier Tocris Bioscience™ Supplier No. 7170/100U
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

TRPC1/6 and Piezo2 inhibitor; resistant to proteolytic digestion

D-GsMTx4 is a TRPC1/6 and Piezo2 inhibitor. Exhibits same effect as L-enantiomer, GsMTx4 (Cat. No. 4912) on TRPC channels. Inhibits mechanosensitive currents by ∼70%. Protects against myocardial infarction in mouse ischemia/reperfusion model. Resistant to proteolytic digestion.

Specifications

Purity 95%
Content And Storage Store at -20°C
Quantity 100 μg
Target Entry TRPC Channel Blockers
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.