Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-(-)-Gulono-γ-Lactone, 99.6%, For HPLC analysis, MP Biomedicals™

Catalog No. ICN15121705 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15121705 5 g
ICN15121710 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15121705 Supplier MP Biomedicals, Inc Supplier No. 0215121705

Chemical Identifiers

CAS 6322-07-2
Molecular Formula C6H10O6
Molecular Weight (g/mol) 178.14
InChI Key SXZYCXMUPBBULW-UHFFFAOYNA-N
Synonym d-gulono-1,4-lactone, d---gulonic acid gamma-lactone, gulonolactone, d-gulonic acid gamma-lactone, gulono-1,4-lactone, d-gulonic acid-1,4-lactone, 3r,4s,5s-5-r-1,2-dihydroxyethyl-3,4-dihydroxydihydrofuran-2 3h-one, 3r,4s,5s-5-1r-1,2-dihydroxyethyl-3,4-dihydroxyoxolan-2-one, d-gulono-gamma-lactone, gulonic acid-1,4-lactone
PubChem CID 165105
IUPAC Name (3R,4S,5S)-5-[(1R)-1,2-dihydroxyethyl]-3,4-dihydroxyoxolan-2-one
SMILES C(C(C1C(C(C(=O)O1)O)O)O)O

Specifications

Color White
Assay Percent Range 99.6%
Quantity 5 g
Formula Weight 178.1
Physical Form Crystalline Powder
Chemical Name or Material D-(-)-Gulono-γ-Lactone, For HPLC analysis
EINECSNumber 228-678-5
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.