Learn More
Lonza Walkersville D-HBE-CF – Diseased Human Bronchial/Tracheal Epithelial Cells – Cystic Fibrosis
Supplier:  Lonza Walkersville 00196979
Encompass
Cystic fibrosis is a disease that causes thick, sticky mucus to build up in the lungs, digestive tract, and other areas of the body. It is one of the most common chronic lung diseases in children and young adults. The disease is the most common, deadly, inherited disorder affecting Caucasians in the United States. The disease eventually worsens to the point where the person is disabled. Today, the average life span for people with CF who live to adulthood is approximately 37 years, a dramatic increase over the last three decades.Benefits:Test negative for mycoplasma, bacteria, yeast, and fungiHIV-1, Hepatitis B and Hepatitis C are not detected for all donors and/or cell lotsA Certificate ofAnalysis is provided for each cell lot purchased Content:1 x Cryopreserved ampule of D-HBE-CF containing ≥500,000 cells
Catalog No. NC2059321