Learn More
Lonza Walkersville D-HBE-COPD Cells - COPD
Catalog No. 5000496287
Encompass
COPD Airway Cells include lung fibroblasts bronchial epithelial and bronchial smooth muscle cells Bronchial epithelial cells from COPD donors are isolated from epithelial lining of airways above bifurcation of the lungs Cryopreserved Diseased Human Bronchial Epithelial cells from COPD donors test negative for mycoplasma bacteria yeast and fungi HIV-1 hepatitis B and hepatitis C are not detected for all donors and/or cell lots A Certificate of Analysis is provided for each cell lot purchased Our catalog number 00195275S are suitable for air-liquid interface studies and are guaranteed to promote full differentiation of the airway epithelium expressed by the formation of a polarized epithelium with good barrier function (transepithelial resistance) secretory phenotype (mucin secretion) and ciliogenesis
Catalog No. 50-004-96287 Supplier Lonza Walkersville Supplier No. 00195275
COPD Airway Cells include lung fibroblasts bronchial epithelial and bronchial smooth muscle cells Bronchial epithelial cells from COPD donors are isolated from epithelial lining of airways above bifurcation of the lungs Cryopreserved Diseased Human Bronchial Epithelial cells from COPD donors test negative for mycoplasma bacteria yeast and fungi HIV-1 hepatitis B and hepatitis C are not detected for all donors and/or cell lots A Certificate of Analysis is provided for each cell lot purchased Our catalog number 00195275S are suitable for air-liquid interface studies and are guaranteed to promote full differentiation of the airway epithelium expressed by the formation of a polarized epithelium with good barrier function (transepithelial resistance) secretory phenotype (mucin secretion) and ciliogenesis