Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Histidine 99.0+%, TCI America™
SDP

Catalog No. H099825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H099825G 25 g
H09985G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H099825G Supplier TCI America Supplier No. H099825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 351-50-8
Molecular Formula C6H9N3O2
Molecular Weight (g/mol) 155.16
MDL Number MFCD00065963
InChI Key HNDVDQJCIGZPNO-RXMQYKEDSA-N
Synonym d-histidine, h-d-his-oh, d-histidin, r-2-amino-3-1h-imidazol-4-yl propanoic acid, d-his, 2r-2-amino-3-1h-imidazol-4-yl propanoic acid, r-2-amino-3-4-imidazolyl propionic acid, 2r-2-amino-3-1h-imidazol-5-yl propanoic acid, r-alpha-amino-1h-imidazole-4-propionic acid, d-alpha-amino-beta-4-imidazolyl propionic acid
PubChem CID 71083
ChEBI CHEBI:27947
IUPAC Name (2R)-2-amino-3-(1H-imidazol-5-yl)propanoic acid
SMILES N[C@H](CC1=CN=CN1)C(O)=O

Specifications

Color White
Quantity 25 g
Formula Weight 155.16
Percent Purity ≥99.0% (T,HPLC)
Physical Form Crystalline Powder
Chemical Name or Material D-Histidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.