Learn More
Lonza Walkersville D-HLF-COPD Fibroblasts - COPD
Catalog No. 5000496218
Encompass
Diseased fibroblasts are isolated from adult COPD donors Cryopreserved Diseased Lung Fibroblast cells stain negative for von Willebrand factor VIII cytokeratins 18 and 19 and alpha smooth muscle actin All cells test negative for mycoplasma bacteria yeast and fungi HIV-1 hepatitis B and hepatitis C are not detected for all donors and/or cell lots A Certificate of Analysis is provided for each cell lot purchased
Catalog No. 50-004-96218 Supplier Lonza Walkersville Supplier No. 00195277
Diseased fibroblasts are isolated from adult COPD donors Cryopreserved Diseased Lung Fibroblast cells stain negative for von Willebrand factor VIII cytokeratins 18 and 19 and alpha smooth muscle actin All cells test negative for mycoplasma bacteria yeast and fungi HIV-1 hepatitis B and hepatitis C are not detected for all donors and/or cell lots A Certificate of Analysis is provided for each cell lot purchased