Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Homoserine 96.0+%, TCI America™
SDP

Catalog No. H12245G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H12245G 5 g
H12241G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H12245G Supplier TCI America Supplier No. H12245G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6027-21-0
Molecular Formula C4H9NO3
Molecular Weight (g/mol) 119.12
MDL Number MFCD00077786
InChI Key UKAUYVFTDYCKQA-GSVOUGTGSA-N
Synonym d-homoserine, 2r-2-amino-4-hydroxybutanoic acid, r-2-amino-4-hydroxybutanoic acid, r-+-2-amino-4-hydroxybutyric acid, h-d-hoser-oh, homoserine, d-homoserin, homoserine, d, ambotzhaa1028, pubchem13826
PubChem CID 2724170
ChEBI CHEBI:30654
IUPAC Name (2R)-2-amino-4-hydroxybutanoic acid
SMILES C(CO)C(C(=O)O)N

Specifications

Melting Point 204°C
Color White-Yellow
Quantity 5 g
Formula Weight 119.12
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material D-Homoserine
EINECSNumber (2)-2759
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.