Learn More
Lonza Walkersville D-HPAEC - Diseased Hu Pulmonar
Supplier:  Lonza Walkersville CC2923
Encompass
Diseased human pulmonary artery endothelial cells are isolated from human donors that have been diagnosed with diabetes type I or type II disease Diseased cells are grown in the same optimized media system as normal human endothelial cells Diabetic cultures can be used for experimental applications in diabetic-related cardiovascular pharmaceutical development vascular pathology including atherosclerosis Cryopreserved Diabetic Type 1 Human Pulmonary Artery Endothelial Cells are shipped in third passage Proliferating Clonetics cells are shipped in fourth passage All cells test negative for mycoplasma bacteria yeast and fungi HIV-1 hepatitis B and hepatitis C are not detected for all donors and/or cell lots A Certificate of Analysis is provided for each cell lot purchased
Catalog No. 50-004-96329