Learn More
Lonza Walkersville D-HPBMC - Diseased - Psoriasis
Catalog No. 5000496262
Encompass
Lonza offers dermal fibroblasts isolated from both lesion sites and non-lesion sites all isolated from the same donor diagnosed with psoriasis Donor Information 69 year old Caucasian male Disease Psoriasis (Lesion Site Derived) Quality Control Testing Seeding Efficiency (26%) Cell Count (582 000 cells/vial) Viability (97%) doubling time (51 hours) HIV/HBV HCV/Mycoplasma (not detected) Sterility (negative) This product is classified as a Limited Edition Cell Type Lonza CellBio Services occasionally obtains excess or additional cells from custom isolations that can be offered as individual cryopreserved vials These Limited Edition Cells are available in limited inventory and may not be available again after inventory is depleted
Catalog No. 50-004-96262 Supplier Lonza Walkersville Supplier No. 00219738
Lonza offers dermal fibroblasts isolated from both lesion sites and non-lesion sites all isolated from the same donor diagnosed with psoriasis Donor Information 69 year old Caucasian male Disease Psoriasis (Lesion Site Derived) Quality Control Testing Seeding Efficiency (26%) Cell Count (582 000 cells/vial) Viability (97%) doubling time (51 hours) HIV/HBV HCV/Mycoplasma (not detected) Sterility (negative) This product is classified as a Limited Edition Cell Type Lonza CellBio Services occasionally obtains excess or additional cells from custom isolations that can be offered as individual cryopreserved vials These Limited Edition Cells are available in limited inventory and may not be available again after inventory is depleted