Learn More
Medchemexpress LLC D-α-Hydroxyglutaric acid | 13095-47-1 | 148.11 | 10MG
SDP
Catalog No. 5000438716
Encompass_Preferred
D-α-Hydroxyglutaric acid is the primary metabolite accumulating in D-2-hydroxyglutaric aciduria, a neurometabolic disease. It acts as a weak competitive antagonist of α-ketoglutarate (α-KG) and inhibits multiple α-KG-dependent dioxygenases. This compound also enhances reactive oxygen species (ROS) production, binds to and inhibits ATP synthase, and suppresses mTOR signaling. It accumulates in human cancers with specific mutations in IDH1 and IDH2 genes.
  • Primary metabolite in D-2-hydroxyglutaric aciduria
  • Weak competitive antagonist of α-ketoglutarate (α-KG)
  • Inhibits multiple α-KG-dependent dioxygenases
  • Increases reactive oxygen species (ROS) production
  • Binds and inhibits ATP synthase
  • Inhibits mTOR signaling
  • Accumulates in human cancers with IDH1 and IDH2 mutations
Catalog No. 50-004-38716 Supplier Medchemexpress LLC Supplier No. HY11303810MG
D-α-Hydroxyglutaric acid is the primary metabolite accumulating in D-2-hydroxyglutaric aciduria, a neurometabolic disease. It acts as a weak competitive antagonist of α-ketoglutarate (α-KG) and inhibits multiple α-KG-dependent dioxygenases. This compound also enhances reactive oxygen species (ROS) production, binds to and inhibits ATP synthase, and suppresses mTOR signaling. It accumulates in human cancers with specific mutations in IDH1 and IDH2 genes.
  • Primary metabolite in D-2-hydroxyglutaric aciduria
  • Weak competitive antagonist of α-ketoglutarate (α-KG)
  • Inhibits multiple α-KG-dependent dioxygenases
  • Increases reactive oxygen species (ROS) production
  • Binds and inhibits ATP synthase
  • Inhibits mTOR signaling
  • Accumulates in human cancers with IDH1 and IDH2 mutations