Learn More
Medchemexpress LLC D-α-Hydroxyglutaric acid | 13095-47-1 | 98.0% | 148.11 | 1 ML
SDP
Catalog No. 5000439795
Encompass_Preferred
D-α-Hydroxyglutaric acid is the primary metabolite that accumulates in the neurometabolic disorder D-2-hydroxyglutaric aciduria. It acts as a weak competitive antagonist of α-ketoglutarate (α-KG), inhibiting multiple α-KG-dependent dioxygenases with a Ki of 10.87 mM. This compound increases reactive oxygen species (ROS) production, binds to and inhibits ATP synthase, and suppresses mTOR signaling. It also accumulates in human cancers with neomorphic mutations in IDH1 and IDH2 genes.
  • Weak competitive antagonist of α-ketoglutarate (α-KG).
  • Inhibits multiple α-KG-dependent dioxygenases.
  • Increases reactive oxygen species (ROS) production.
  • Binds to and inhibits ATP synthase.
  • Suppresses mTOR signaling.
  • Accumulates in human cancers with IDH1 and IDH2 gene mutations.
Catalog No. 50-004-39795 Supplier Medchemexpress LLC Supplier No. HY11303810MM/1ML
D-α-Hydroxyglutaric acid is the primary metabolite that accumulates in the neurometabolic disorder D-2-hydroxyglutaric aciduria. It acts as a weak competitive antagonist of α-ketoglutarate (α-KG), inhibiting multiple α-KG-dependent dioxygenases with a Ki of 10.87 mM. This compound increases reactive oxygen species (ROS) production, binds to and inhibits ATP synthase, and suppresses mTOR signaling. It also accumulates in human cancers with neomorphic mutations in IDH1 and IDH2 genes.
  • Weak competitive antagonist of α-ketoglutarate (α-KG).
  • Inhibits multiple α-KG-dependent dioxygenases.
  • Increases reactive oxygen species (ROS) production.
  • Binds to and inhibits ATP synthase.
  • Suppresses mTOR signaling.
  • Accumulates in human cancers with IDH1 and IDH2 gene mutations.