Learn More
Medchemexpress LLC D-α-hydroxyglutaric acid disodium | 103404-90-6 | 98.0% | 192.08 | C5H6Na2O5 | 10 MG
SDP
Catalog No. 5000212352
Encompass_Preferred
D-α-hydroxyglutaric acid disodium is the disodium salt of the R-enantiomer of 2-hydroxyglutaric acid, supplied as a solid research reagent and biochemical standard for studies of D-2-hydroxyglutaric acid metabolism and α-ketoglutarate-dependent enzyme activity. Typical properties: C5H6Na2O5, Mw 192.08, CAS 103404-90-6.
  • High purity suitable for research applications (>98%).
  • Disodium salt form for improved aqueous solubility.
  • Useful as a biochemical standard in metabolic and enzyme studies.
  • Reported to modulate ATP synthase activity and increase ROS in biochemical assays.
  • Stable when stored sealed away from moisture; in solvent: -80°C (6 months), -20°C (1 month).
Catalog No. 50-002-12352 Supplier Medchemexpress LLC Supplier No. HY10054210MG
D-α-hydroxyglutaric acid disodium is the disodium salt of the R-enantiomer of 2-hydroxyglutaric acid, supplied as a solid research reagent and biochemical standard for studies of D-2-hydroxyglutaric acid metabolism and α-ketoglutarate-dependent enzyme activity. Typical properties: C5H6Na2O5, Mw 192.08, CAS 103404-90-6.
  • High purity suitable for research applications (>98%).
  • Disodium salt form for improved aqueous solubility.
  • Useful as a biochemical standard in metabolic and enzyme studies.
  • Reported to modulate ATP synthase activity and increase ROS in biochemical assays.
  • Stable when stored sealed away from moisture; in solvent: -80°C (6 months), -20°C (1 month).