Learn More
Medchemexpress LLC D-α-hydroxyglutaric acid disodium | 103404-90-6 | MFCD00069573 | >95.0% | 192.08 | C5H6Na2O5 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10054225MG
Encompass_Preferred
D-α-hydroxyglutaric acid disodium is the disodium salt of the R-enantiomer of 2-hydroxyglutarate, provided as a research reagent for biochemical and analytical studies. It is the principal metabolite in D-2-hydroxyglutaric aciduria and acts as a weak competitive antagonist of α-ketoglutarate, affecting α-KG-dependent dioxygenases. Molecular formula C5H6Na2O5; molecular weight 192.08; store at 4°C, sealed and away from moisture.
  • Disodium salt form for improved solubility and handling.
  • R-enantiomer specificity for enantioselective biological studies.
  • Inhibits α-KG-dependent dioxygenases for mechanistic assays.
  • Useful for biochemical, metabolic, and analytical research applications.
  • Known molecular formula and molecular weight for formulation and dosing calculations.
  • Stable when stored sealed at 4°C away from moisture.
Catalog No. 50-002-12785