Learn More
Medchemexpress LLC D-α-hydroxyglutaric acid disodium | 103404-90-6 | MFCD00069573 | 98.0% | 192.08 g/mol | C5H6Na2O5 | 1 ML
SDP
Catalog No. 5000212669
Encompass_Preferred
D-α-hydroxyglutaric acid disodium is the disodium salt of the D-enantiomer of 2-hydroxyglutaric acid, provided as a 10 mM solution (1 mL) or as solids. It is used as a biochemical research reagent to probe α-ketoglutarate-dependent enzymes, metabolic and mitochondrial function, and oncometabolite biology. Purity is typically 98.0%.
  • Available as 10 mM solution in water (1 mL) or as solid quantities for flexibility.
  • Useful for studies of α-ketoglutarate-dependent dioxygenases and epigenetic modifiers.
  • Reported to increase reactive oxygen species and inhibit ATP synthase and mTOR signaling.
  • High purity (98.0%) suitable for biochemical and cellular assays.
  • Documents provided for quality and safety: data sheet, COA, and SDS.
Catalog No. 50-002-12669 Supplier Medchemexpress LLC Supplier No. HY10054210MM 1ML
D-α-hydroxyglutaric acid disodium is the disodium salt of the D-enantiomer of 2-hydroxyglutaric acid, provided as a 10 mM solution (1 mL) or as solids. It is used as a biochemical research reagent to probe α-ketoglutarate-dependent enzymes, metabolic and mitochondrial function, and oncometabolite biology. Purity is typically 98.0%.
  • Available as 10 mM solution in water (1 mL) or as solid quantities for flexibility.
  • Useful for studies of α-ketoglutarate-dependent dioxygenases and epigenetic modifiers.
  • Reported to increase reactive oxygen species and inhibit ATP synthase and mTOR signaling.
  • High purity (98.0%) suitable for biochemical and cellular assays.
  • Documents provided for quality and safety: data sheet, COA, and SDS.