Learn More
Medchemexpress LLC D-α-hydroxyglutaric acid disodium | 103404-90-6 | MFCD00069573 | 98.0% | 192.08 g/mol | C5H6Na2O5 | 5 MG
SDP
Catalog No. 5000211909
Encompass_Preferred
D-α-Hydroxyglutaric acid disodium is the disodium salt of the D-enantiomer of 2-hydroxyglutaric acid used as a biochemical research reagent. It acts as a weak competitive antagonist of α-ketoglutarate, inhibits multiple α-KG-dependent dioxygenases (Ki ≈ 10.87 mM), increases reactive oxygen species production, binds and inhibits ATP synthase, and modulates mTOR signaling. Supplied in both microgram/milligram quantities and as a ready-to-use aqueous solution for laboratory experiments.
  • Useful in studies of α-KG-dependent dioxygenase activity.
  • Modulates mitochondrial function by inhibiting ATP synthase.
  • Induces reactive oxygen species for oxidative stress assays.
  • Available as small mass packs and as a 10 mM solution for convenience.
  • Molecular weight 192.08 g/mol for accurate dosing and calculations.
Catalog No. 50-002-11909 Supplier Medchemexpress LLC Supplier No. HY1005425MG
D-α-Hydroxyglutaric acid disodium is the disodium salt of the D-enantiomer of 2-hydroxyglutaric acid used as a biochemical research reagent. It acts as a weak competitive antagonist of α-ketoglutarate, inhibits multiple α-KG-dependent dioxygenases (Ki ≈ 10.87 mM), increases reactive oxygen species production, binds and inhibits ATP synthase, and modulates mTOR signaling. Supplied in both microgram/milligram quantities and as a ready-to-use aqueous solution for laboratory experiments.
  • Useful in studies of α-KG-dependent dioxygenase activity.
  • Modulates mitochondrial function by inhibiting ATP synthase.
  • Induces reactive oxygen species for oxidative stress assays.
  • Available as small mass packs and as a 10 mM solution for convenience.
  • Molecular weight 192.08 g/mol for accurate dosing and calculations.