Learn More
Medchemexpress LLC HY-124691 5mg Medchemexpress, D-I03 CAS:688342-78-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1246915MG
Encompass_Preferred
Medchemexpress, HY-124691 5mg D-I03 CAS:688342-78-1 D-I03 is a selective RAD52 inhibitor with a Kd of 25.8 µM. D-I03 specifically inhibits RAD52-dependent single-strand annealing (SSA) and D-loop formation with IC50s of 5 µM and 8 µM, respectively. D-I03 suppresses growth of BRCA1- and BRCA2-deficient cells and inhibits formation of damage-induced RAD52 foci, but does not effect on RAD51 foci induced by Cisplatin. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9677