Learn More
AdipoGen D-Iditol
SDP
Supplier:  AdipoGen CDXD0854M100
Encompass_Preferred
Chemical. CAS 25878-23-3. Formula C6H14O6. Molecular Weight 182.17. D-Iditol is a chiral molecule for chemical synthesis. It has been shown to inhibit glucosidase I, but not glucosidase II when used at a concentration of 1 mM.
Catalog No. 50-256-1033