Learn More
Medchemexpress LLC D-Iditol | 25878-23-3 | MFCD09951922 | 98.0% | 182.17 g/mol | C6H14O6 | 10 MG
SDP
Catalog No. 5000283841
Encompass_Preferred
D-Iditol is a fungal metabolite and sugar alcohol that accumulates in galactokinase deficiency. It is supplied as a white to off-white solid with ≥98.0% purity for research use only and is commonly used as an endogenous metabolite reference or reagent in biochemical and metabolic studies.
  • High purity: 98.0% (manufacturer specification).
  • Molecular weight 182.17 g/mol; formula C6H14O6.
  • White to off-white solid suitable for analytical and synthetic applications.
  • Stable when stored sealed at 4°C; in solvent: -80°C (6 months) or -20°C (1 month).
  • Suitable for biochemical, metabolic, and analytical research workflows.
Catalog No. 50-002-83841 Supplier Medchemexpress LLC Supplier No. HYW14558410MG
D-Iditol is a fungal metabolite and sugar alcohol that accumulates in galactokinase deficiency. It is supplied as a white to off-white solid with ≥98.0% purity for research use only and is commonly used as an endogenous metabolite reference or reagent in biochemical and metabolic studies.
  • High purity: 98.0% (manufacturer specification).
  • Molecular weight 182.17 g/mol; formula C6H14O6.
  • White to off-white solid suitable for analytical and synthetic applications.
  • Stable when stored sealed at 4°C; in solvent: -80°C (6 months) or -20°C (1 month).
  • Suitable for biochemical, metabolic, and analytical research workflows.