Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-Isovaline | 3059-97-0 | 97.0% | 117.15 | 5 G
SDP

Catalog No. 5000395586
Encompass_Preferred

D-Isovaline is a valine derivative, useful in various applications as an amino acid derivative. These compounds are recognized as beneficial ergogenic dietary substances, influencing anabolic hormone secretion, fuel supply during exercise, and mental performance under stress.

  • Solid appearance
  • White to off-white color
  • Purity of 97.0%
  • Molecular weight of 117.15
  • Consistent with structure by 1H NMR Spectrum

Catalog No. 50-003-95586 Supplier Medchemexpress LLC Supplier No. HYW0085445G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Isovaline is a valine derivative, useful in various applications as an amino acid derivative. These compounds are recognized as beneficial ergogenic dietary substances, influencing anabolic hormone secretion, fuel supply during exercise, and mental performance under stress.

  • Solid appearance
  • White to off-white color
  • Purity of 97.0%
  • Molecular weight of 117.15
  • Consistent with structure by 1H NMR Spectrum

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.