Learn More
Medchemexpress LLC HY-P0069 5mg , D-JNKI-1 CAS:1445179-97-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP00695MG
Encompass_Preferred
HY-P0069 5mg D-JNKI-1 CAS: 1445179-97-4 D-JNKI-1 (AM-111) is a highly potent and cell-permeable peptide inhibitor of JNK. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1198