Learn More
Medchemexpress LLC d-KLA Peptide | 286380-05-0 | 99.3% | 1523.99 | 1 MG
SDP
Catalog No. 5000425755
Encompass_Preferred
d-KLA Peptide is a synthetic pro-apoptotic peptide that specifically targets mitochondria and induces apoptosis by destroying the mitochondrial membrane. It activates biochemical pathways associated with apoptosis, including the activation of caspase family proteins and PARP (poly ADP ribose polymerase). d-KLA Peptide can be used to carry and deliver genes or small molecules to enhance anti-tumor effects.
  • Specific targeting of mitochondria to induce apoptosis
  • Destroys the mitochondrial membrane
  • Activates caspase family proteins and PARP
  • Can enhance anti-tumor effects when used to carry and deliver genes or small molecules
Catalog No. 50-004-25755 Supplier Medchemexpress LLC Supplier No. HYP102851MG
d-KLA Peptide is a synthetic pro-apoptotic peptide that specifically targets mitochondria and induces apoptosis by destroying the mitochondrial membrane. It activates biochemical pathways associated with apoptosis, including the activation of caspase family proteins and PARP (poly ADP ribose polymerase). d-KLA Peptide can be used to carry and deliver genes or small molecules to enhance anti-tumor effects.
  • Specific targeting of mitochondria to induce apoptosis
  • Destroys the mitochondrial membrane
  • Activates caspase family proteins and PARP
  • Can enhance anti-tumor effects when used to carry and deliver genes or small molecules